MilliporeSigma
  • IL-15 regulates Bcl-2 family members Bim and Mcl-1 through JAK/STAT and PI3K/AKT pathways in T cells.
IL-15 regulates Bcl-2 family members Bim and Mcl-1 through JAK/STAT and PI3K/AKT pathways in T cells.
European journal of immunology (2014-05-16)
Aparna R Shenoy, Susanne Kirschnek, Georg Häcker
ABSTRACT
Maintenance of T cells is determined by their survival capacity, which is regulated by Bcl-2 proteins. Cytokines signalling through the common gamma chains such as IL-2, IL-7 and IL-15 are important for T-cell survival but how these cytokines determine the expression of Bcl-2-family proteins is not clear. We report signalling events of cytokines that regulate expression of two key Bcl-2 proteins, pro-apoptotic Bim and anti-apoptotic Mcl-1, in resting C57BL/6 mouse T cells. IL-2, IL-7 and IL-15 inhibited apoptosis but paradoxically induced the expression of Bim, countered by concomitant induction of Mcl-1. Bim induction by IL-15 was found at the mRNA and protein levels and depended on both JAK/STAT and PI3K signals. A new STAT5-binding site was identified in the Bim promoter, which was occupied by STAT5 upon IL-15 stimulation. Although it also depended on JAK/STAT- and PI3K signalling, Mcl-1 regulation was independent of Mcl-1 mRNA levels and of regulation of protein stability, suggesting translational regulation. Concurrent CD3 signals inhibited some of the IL-7 effect but not the IL-15 effect on Bcl-2 proteins. The data suggest that cytokines induce Bim and prime T cells for apoptosis, but also inhibit apoptosis by stabilising Mcl-1. Later downregulation of short-lived Mcl-1 may induce efficient, Bim-dependent apoptosis.
Product Description
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Interleukin-2 from mouse, IL-2, recombinant, expressed in E. coli, carrier free
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)