MilliporeSigma
  • Rab40C is a novel Varp-binding protein that promotes proteasomal degradation of Varp in melanocytes.
Rab40C is a novel Varp-binding protein that promotes proteasomal degradation of Varp in melanocytes.
Biology open (2015-02-11)
Ayaka Yatsu, Hikaru Shimada, Norihiko Ohbayashi, Mitsunori Fukuda
ABSTRACT
Varp (VPS9-ankyrin repeat protein) was originally identified as an activator of small GTPase Rab21 through its VPS9 domain, but it has subsequently been shown to function as a Rab32/38 effector through its first ANKR1 domain. Although these functions of Varp are important for melanogenesis, Varp contains a second ANKR2 domain, whose function remained completely unknown. Here we identified Rab40C, an atypical Rab containing a SOCS box that recruits a ubiquitin ligase complex, as a novel ANKR2-binding protein and investigated its involvement in melanogenic enzyme trafficking in melanocytes. The results showed that overexpression of Rab40C in melanocytes caused a dramatic reduction in melanogenic enzyme Tyrp1 signals by promoting proteasomal degradation of Varp in a SOCS-box-dependent manner and that knockdown of Rab40C in melanocytes caused an increase in the amount of Varp. Intriguingly, Rab40C knockdown also caused a dramatic reduction in Tyrp1 signals, the same as Varp overexpression did. These findings indicated that Rab40C is a previously unexpected regulator of Tyrp1 trafficking in melanocytes through controlling the proteasomal degradation of Varp.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
N-Ethylmaleimide, purum p.a., ≥99.0% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
ANTI-FLAG® M2-Peroxidase (HRP) antibody, Mouse monoclonal, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Roche
Anti-HA-Peroxidase, High Affinity, from rat IgG1
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.