MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Evidence for respiratory neuromodulator interdependence after cholinergic disruption in the ventral respiratory column.
Evidence for respiratory neuromodulator interdependence after cholinergic disruption in the ventral respiratory column.
Respiratory physiology & neurobiology (2014-09-30)
Clarissa Muere, Suzanne Neumueller, Justin Miller, Samantha Olesiak, Matthew R Hodges, Lawrence Pan, Hubert V Forster
ABSTRACT
Reverse dialysis of the muscarinic receptor antagonist, atropine (ATR, 50 mM), into the pre-Bötzinger Complex region of the ventral respiratory column (VRC) of awake and sleeping goats increases breathing frequency and serotonin (5-HT), substance P (SP), glycine, and GABA concentrations in the effluent dialysate. Herein, we report data from goats in which we reverse dialyzed 5 mM ATR or specific antagonists of M2 or M3 muscarinic receptors into the VRC. The effects on frequency of all three antagonists were not significantly different from time control studies. 5 mM ATR and the M3 antagonist increased SP sevenfold less than 50 mM ATR. The antagonists had no effect on 5-HT, glycine, and/or GABA, suggesting that the increases in glycine and GABA with 50 mM ATR were secondary to the larger increases in 5-HT and/or SP. These data are suggestive of neuromodulator interdependence, whereby attenuation of one neuromodulator is compensated for by local changes in other neuromodulators to stabilize breathing.
Sigma-Aldrich
Calcium chloride solution, 0.025 M
Sigma-Aldrich
Calcium chloride solution, 3.2 mM
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
4-DAMP, ≥98% (HPLC)
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Methoctramine hydrate, ≥97% (NMR), solid
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%