MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • MiR-21 promotes intrahepatic cholangiocarcinoma proliferation and growth in vitro and in vivo by targeting PTPN14 and PTEN.
MiR-21 promotes intrahepatic cholangiocarcinoma proliferation and growth in vitro and in vivo by targeting PTPN14 and PTEN.
Oncotarget (2015-03-25)
Li-Juan Wang, Chen-Chen He, Xin Sui, Meng-Jiao Cai, Cong-Ya Zhou, Jin-Lu Ma, Lei Wu, Hao Wang, Su-Xia Han, Qing Zhu
ABSTRACT
Intrahepatic cholangiocarcinoma (ICC) constitutes the second-most common primary hepatic malignancy. MicroRNAs (miRNAs) play important roles in the pathogenesis of ICC. However, the clinical significance of miR-21 levels in ICC remains unclear. Here, we investigated the role of miR-21 in ICC and found that its expression was significantly upregulated in serum of ICC patients. Serum miR-21 levels robustly distinguished ICC patients from control subjects. Further experiments showed that inhibition of miR-21 suppressed ICC cell proliferation in vitro and tumor growth in vivo. Specifically, inhibition of miR-21 induced cell cycle arrest and apoptosis. Moreover, PTPN14 and PTEN were identified as direct and functional targets of miR-21. Finally, we showed high expression levels of miR-21 were closely related to adverse clinical features, diminished survival, and poor prognosis in ICC patients. This study revealed functional and mechanistic links between miR-21 and tumor suppressor genes, PTPN14 and PTEN, in the pathogenesis of ICC. MiR-21 not only plays important roles in the regulation of cell proliferation and tumor growth in ICC, but is also a diagnostic and prognostic marker, and a potential therapeutic target for ICC.
Product Description
Supelco
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Sigma-Aldrich
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
SAFC
Hexadecyltrimethylammonium bromide, USP/NF
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Supelco
Digoxigenin, analytical standard
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Formamide, ACS reagent, ≥99.5%
Sigma-Aldrich
Formamide, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Formamide, ≥99.5% (GC), BioReagent, Molecular Biology
Sigma-Aldrich
Formamide, JIS special grade, ≥98.5%
Sigma-Aldrich
Formamide, SAJ first grade, ≥98.5%