MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • The spino-bulbar-cerebellar pathway: Activation of neurons projecting to the lateral reticular nucleus in the rat in response to noxious mechanical stimuli.
The spino-bulbar-cerebellar pathway: Activation of neurons projecting to the lateral reticular nucleus in the rat in response to noxious mechanical stimuli.
Neuroscience letters (2015-02-26)
Zilli Huma, Kirsty Ireland, David J Maxwell
ABSTRACT
It is now well established that the cerebellum receives input from nociceptors which may serve to adjust motor programmes in response to pain and injury. In this study, we investigated the possibility that spinoreticular neurons (SRT) which project to a pre-cerebellar nucleus, the lateral reticular nucleus (LRt), respond to noxious mechanical stimulation. Seven adult male rats received stereotaxic injections of the b subunit of cholera toxin in the LRt. Following a 5 day interval, animals were anesthetised with urethane and a noxious mechanical stimulus was applied to the right hind paw. Animals were fixed by perfusion 5min following application of the stimulus. Retrogradely labelled SRT neurons of the lumbar spinal cord were examined for immunoreactivity for phosphorylated ERK (pERK) and the neurokinin-1 (NK-1) receptor. Approximately 15% of SRT cells in deep laminae (IV-VII and X) expressed pERK ipsilateral to the site of the stimulus. Around 60% of SRT cells with the NK-1 receptor expressed pERK but 5% of pERK expressing cells were negatively labelled for NK-1. It is concluded that a significant proportion of SRT cells projecting to the LRt respond to noxious mechanical stimuli and that one of the functions of this pathway may be to provide the cerebellum with nociceptive information.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Supelco
Meprobamate solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Carprofen for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Meprobamate, analytical standard
Supelco
Carprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Carprofen, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Supelco
Carprofen, VETRANAL®, analytical standard