MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Prevention of biofilm formation by methacrylate-based copolymer films loaded with rifampin, clarithromycin, doxycycline alone or in combination.
Prevention of biofilm formation by methacrylate-based copolymer films loaded with rifampin, clarithromycin, doxycycline alone or in combination.
Pharmaceutical research (2014-06-18)
Warren E Rose, Daniel P Otto, Marique E Aucamp, Zach Miller, Melgardt M de Villiers
ABSTRACT
This study reports the incorporation of the antibiotics rifampin, doxycycline and clarithromycin in poly(styrene-co-methyl methacrylate films and their effect on biofilm prevention. Invasive procedures in patients such as surgical device, or intravenous or urinary catheter implantation, often results in complicated hospital-acquired nosocomial infections. Biofilm formation is essential to establish these infections on these devices and novel antibiotic delivery approaches are needed for more effective management. The films were evaluated in vitro for drug release and for their ability to prevent biofilm formation by methicillin susceptible and methicillin resistant Staphylococcus aureus. Surface tension components, obtained from contact angle measurements, and the morphology of the films evaluated by scanning electron microscopy were also investigated. In this study, antibiotic-loaded methacrylic copolymer films that effectively released rifampin, clarithromycin and doxycycline for up to 21 days prevented biofilm formation when tested in an in vitro bioreactor model. These drug loaded copolymer films provided the advantage by coating materials with a novel surface that was unsuitable for resettling of biofilms once the antibiotic was dissolved from the polymer surface. A combination of rifampin and clarithromycin released from the polymer film provided >99.9% kill of an MRSA inoculate for up to 72 h. Results showed that combining multiple drugs in copolymer films with unique surface properties, initial hydrophilicity and increase in roughness, can be an effective way to prevent biofilm formation.
Sigma-Aldrich
3,3′-Diindolylmethane, ≥98% (HPLC)
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
Aluminum oxide, CP
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminum oxide, Brockmann I, activated, standard grade, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, standard grade, Brockmann I, activated, basic
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, pellets, 3 mm
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, Brockmann I, standard grade, activated, acidic
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, Brockmann I, standard grade, activated, neutral, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, activated, Brockmann I, standard grade, neutral
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm