MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effect of a dienogest for an experimental three-dimensional endometrial culture model for endometriosis.
Effect of a dienogest for an experimental three-dimensional endometrial culture model for endometriosis.
Medical molecular morphology (2013-10-22)
Japarath Prechapanich, Takeshi Kajihara, Keiko Fujita, Kazuko Sato, Satomi Uchino, Kayoko Tanaka, Sachiko Matsumoto, Masumi Akita, Masabumi Nagashima, Jan J Brosens, Osamu Ishihara
ABSTRACT
The pathogenesis of endometriosis remains poorly understood at least in part because early stages of the disease process are difficult to investigate. Previous studies have proposed a three-dimensional fibrin matrix culture model to study human endometriosis. We examined the ultrastructural features of the endometriosis in this model and assessed the effect of a progestin on endometrial outgrowth and apoptosis in this culture system. Endometrial explants were placed in three-dimensional fibrin matrix culture and treated with and without various concentrations of the progestin dienogest. By the second week, endometrial gland-like formation was established in outgrowths both attached to and at a distance from the explants. These cells formed a combination of clumps and tubular monolayers surrounding a central cavity. Electron microscopy demonstrated that these cells are polarized with microvilli on the apical surface, desmosome-like structures, and basement membrane; features consistent with glandular epithelial cells. Outgrowth of endometrial stromal cells and glandular formation was impaired in response to dienogest in a dose-dependent manner. Our study shows that the human endometrial explants cultured in three-dimensional fibrin matrix establish outgrowths that ultrastructurally resemble ectopic endometrial implants. This model may provide insight into the cellular processes leading to endometriosis formation and enables screening of therapeutic compounds.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)