MilliporeSigma
  • Dihydromyricetin activates AMP-activated protein kinase and P38(MAPK) exerting antitumor potential in osteosarcoma.
Dihydromyricetin activates AMP-activated protein kinase and P38(MAPK) exerting antitumor potential in osteosarcoma.
Cancer prevention research (Philadelphia, Pa.) (2014-06-05)
Zhiqiang Zhao, Jun-qiang Yin, Man-si Wu, Guohui Song, Xian-biao Xie, Changye Zou, Qinglian Tang, Yuanzhong Wu, Jinchang Lu, Yongqian Wang, Jin Wang, Tiebang Kang, Qiang Jia, Jingnan Shen
ABSTRACT
Numerous patients with osteosarcoma either are not sensitive to chemotherapy or develop drug resistance to current chemotherapy regimens. Therefore, it is necessary to develop several potentially useful therapeutic agents. Dihydromyricetin is the major flavonoid component derived from Ampelopsis grossedentata, which has a long history of use in food and medicine. The present study examined the antitumor activity both in vitro and in vivo without noticeable side effects and the underlying mechanism of action of dihydromyricetin in osteosarcoma cells. We found that dihydromyricetin induced increased p21 expression and G2-M cell-cycle arrest, caused DNA damage, activated ATM-CHK2-H2AX signaling pathways, and induced apoptosis in osteosarcoma cells as well as decreasing the sphere formation capability by downregulating Sox2 expression. Mechanistic analysis showed that the antitumor potential of dihydromyricetin may be due to the activation of AMPKα and p38(MAPK), as the activating AMPKα led to the inactivation of GSK3β in osteosarcoma cells. Moreover, GSK3β deletion or GSK3β inhibition by LiCl treatment resulted in increased p21 expression and reduced Sox2 expression in osteosarcoma cells. Taken together, our results strongly indicate that the antitumor potential of dihydromyricetin is correlated with P38(MAPK) and the AMPKα-GSK3β-Sox2 signaling pathway. Finally, immunohistochemical analysis indicated that some patients had a lower p-AMPK expression after chemotherapy, which supports that the combination of dihydromyricetin and chemotherapy drug will be beneficial for patients with osteosarcoma. In conclusion, our results are the first to suggest that dihydromyricetin may be a therapeutic candidate for the treatment of osteosarcoma.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri