MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effect of lithium chloride on endoplasmic reticulum stress-related PERK/ROCK signaling in a rat model of glaucoma.
Effect of lithium chloride on endoplasmic reticulum stress-related PERK/ROCK signaling in a rat model of glaucoma.
Die Pharmazie (2015-05-09)
Xiao-Bo Sun, Hong-E Lu, Yuan Chen, Xiao-Hui Fan, Bin Tong
ABSTRACT
Elevated intraocular pressure (IOP) is considered as the major risk factor for the loss of retinal ganglion cells (RGCs) and their axons in glaucoma. Lithium chloride (LiCl) inhibits glycogen synthase kinase-3 beta (GSK-3β) and attends PERK-induced endoplasmic reticulum stress (ERs) transition. PERK is a type I transmembrane protein located in the endoplasmic reticulum. PERK pathway activation takes place in ERs early inhibiting protein synthnesis to protect cell and promote cell survival. Here, we firstly evaluate that LiCl reduced IOP when administered intraperitoneally. After 6 weeks, IOP dropped by around 21.9% in LiCl treated rats. Then we investigated the effects of LiCI on PERK-mediated signaling pathways. LiCl treatment activated PERK and inhibited the expression of ROCK-1 and ROCK-2 in a rat model of glaucoma. Collectively, these results suggest that LiCl reduced the IOP through the phosphorylation of PERK by the regulation of PERK/ROCK signaling in glaucoma rat model.
Product Description
Supelco
Electrolyte solution, nonaqueous, 2 M LiCl in ethanol
Supelco
Lithium chloride solution, 1 M in ethanol
Sigma-Aldrich
Lithium chloride, puriss. p.a., anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium chloride solution, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, Molecular Biology, ≥99%
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Lithium chloride solution, 2 M in ethanol