MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Placental development during early pregnancy in sheep: effects of embryo origin on vascularization.
Placental development during early pregnancy in sheep: effects of embryo origin on vascularization.
Reproduction (Cambridge, England) (2014-01-30)
Anna T Grazul-Bilska, Mary Lynn Johnson, Pawel P Borowicz, Jerzy J Bilski, Taylor Cymbaluk, Spencer Norberg, Dale A Redmer, Lawrence P Reynolds
ABSTRACT
Utero-placental growth and vascular development are critical for pregnancy establishment that may be altered by various factors including assisted reproductive technologies (ART), nutrition, or others, leading to compromised pregnancy. We hypothesized that placental vascularization and expression of angiogenic factors are altered early in pregnancies after transfer of embryos created using selected ART methods. Pregnancies were achieved through natural mating (NAT), or transfer of embryos from NAT (NAT-ET), or IVF or in vitro activation (IVA). Placental tissues were collected on day 22 of pregnancy. In maternal caruncles (CAR), vascular cell proliferation was less (P<0.05) for IVA than other groups. Compared with NAT, density of blood vessels was less (P<0.05) for IVF and IVA in fetal membranes (FM) and for NAT-ET, IVF, and IVA in CAR. In FM, mRNA expression was decreased (P<0.01-0.08) in NAT-ET, IVF, and IVA compared with NAT for vascular endothelial growth factor (VEGF) and its receptor FLT1, placental growth factor (PGF), neuropilin 1 (NP1) and NP2, angiopoietin 1 (ANGPT1) and ANGPT2, endothelial nitric oxide synthase 3 (NOS3), hypoxia-inducible factor 1A (HIF1A), fibroblast growth factor 2 (FGF2), and its receptor FGFR2. In CAR, mRNA expression was decreased (P<0.01-0.05) in NAT-ET, IVF, and IVA compared with NAT for VEGF, FLT1, PGF, ANGPT1, and TEK. Decreased mRNA expression for 12 of 14 angiogenic factors across FM and CAR in NAT-ET, IVF, and IVA pregnancies was associated with reduced placental vascular development, which would lead to poor placental function and compromised fetal and placental growth and development.
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
6-(Dimethylamino)purine, ≥98%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%