MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Near-infrared imaging for high-throughput screening of moisture induced changes in freeze-dried formulations.
Near-infrared imaging for high-throughput screening of moisture induced changes in freeze-dried formulations.
Journal of pharmaceutical sciences (2014-03-26)
Hjalte Trnka, Anna Palou, Pierre Emanuel Panouillot, Ari Kauppinen, Maunu Toiviainen, Holger Grohganz, Manel Alcalà, Mikko Juuti, Jarkko Ketolainen, Jukka Rantanen
ABSTRACT
Evaluation of freeze-dried biopharmaceutical formulations requires careful analysis of multiple quality attributes. The aim of this study was to evaluate the use of near-infrared (NIR) imaging for fast analysis of water content and related physical properties in freeze-dried formulations. Model formulations were freeze-dried in well plates. Samples were imaged with a NIR hyperspectral camera after freeze-drying and upon storage. On the basis of Karl Fischer titration reference values, a univariate quantification model was constructed and used to visualize the distribution of water within freeze-dried samples. Differences observed between samples stored at 11% and 43% relative humidity (RH) were found to be related to the amount of amorphous component in the sample. When stored at 43% RH, the moisture content in samples with high sucrose content increased within 2 days and some degree of localized drying was observed within the samples after 3 days of storage. Further investigations with X-ray powder diffraction confirmed this local drying to be related to crystallization of sucrose. The combination of fast analysis of water content and spatial solid-state information makes NIR imaging a powerful tool for formulation development of freeze-dried samples.
Product Description
Mannitol, European Pharmacopoeia (EP) Reference Standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, SAJ first grade, ≥99.0%
Sigma-Aldrich
D-Mannitol, SAJ special grade, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, JIS special grade
Sigma-Aldrich
Sucrose, SAJ first grade
Sigma-Aldrich
Sucrose, meets USP testing specifications
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture