MilliporeSigma
  • Diagnostic accuracy of pulmonary host inflammatory mediators in the exclusion of ventilator-acquired pneumonia.
Diagnostic accuracy of pulmonary host inflammatory mediators in the exclusion of ventilator-acquired pneumonia.
Thorax (2014-10-10)
Thomas P Hellyer, Andrew Conway Morris, Daniel F McAuley, Timothy S Walsh, Niall H Anderson, Suveer Singh, Paul Dark, Alistair I Roy, Simon V Baudouin, Stephen E Wright, Gavin D Perkins, Kallirroi Kefala, Melinda Jeffels, Ronan McMullan, Cecilia M O'Kane, Craig Spencer, Shondipon Laha, Nicole Robin, Savita Gossain, Kate Gould, Marie-Hélène Ruchaud-Sparagano, Jonathan Scott, Emma M Browne, James G MacFarlane, Sarah Wiscombe, John D Widdrington, Ian Dimmick, Ian F Laurenson, Frans Nauwelaers, A John Simpson
ABSTRACT
Excessive use of empirical antibiotics is common in critically ill patients. Rapid biomarker-based exclusion of infection may improve antibiotic stewardship in ventilator-acquired pneumonia (VAP). However, successful validation of the usefulness of potential markers in this setting is exceptionally rare. We sought to validate the capacity for specific host inflammatory mediators to exclude pneumonia in patients with suspected VAP. A prospective, multicentre, validation study of patients with suspected VAP was conducted in 12 intensive care units. VAP was confirmed following bronchoscopy by culture of a potential pathogen in bronchoalveolar lavage fluid (BALF) at >10(4) colony forming units per millilitre (cfu/mL). Interleukin-1 beta (IL-1β), IL-8, matrix metalloproteinase-8 (MMP-8), MMP-9 and human neutrophil elastase (HNE) were quantified in BALF. Diagnostic utility was determined for biomarkers individually and in combination. Paired BALF culture and biomarker results were available for 150 patients. 53 patients (35%) had VAP and 97 (65%) patients formed the non-VAP group. All biomarkers were significantly higher in the VAP group (p<0.001). The area under the receiver operator characteristic curve for IL-1β was 0.81; IL-8, 0.74; MMP-8, 0.76; MMP-9, 0.79 and HNE, 0.78. A combination of IL-1β and IL-8, at the optimal cut-point, excluded VAP with a sensitivity of 100%, a specificity of 44.3% and a post-test probability of 0% (95% CI 0% to 9.2%). Low BALF IL-1β in combination with IL-8 confidently excludes VAP and could form a rapid biomarker-based rule-out test, with the potential to improve antibiotic stewardship.
Product Description
Supelco
Water, for TOC analysis
Pricing and availability is not currently available.
Supelco
Water, for ultratrace analysis, ACS reagent
Pricing and availability is not currently available.
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pricing and availability is not currently available.
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Pricing and availability is not currently available.
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Pricing and availability is not currently available.
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Water, BioPerformance Certified
Pricing and availability is not currently available.
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Pricing and availability is not currently available.
Supelco
Water, suitable for ion chromatography
Pricing and availability is not currently available.
Sigma-Aldrich
Water, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Pricing and availability is not currently available.
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Water, PCR Reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Pricing and availability is not currently available.
Sigma-Aldrich
Water, HPLC Plus
Pricing and availability is not currently available.
Sigma-Aldrich
Water, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Water, Deionized
Pricing and availability is not currently available.
Sigma-Aldrich
Water, ACS reagent
Pricing and availability is not currently available.
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pricing and availability is not currently available.
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Pricing and availability is not currently available.
Sigma-Aldrich
2,3-Dimercapto-1-propanol, ≥98% (iodometric)
Pricing and availability is not currently available.