MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Influence of dose and animal species on accelerated blood clearance of PEGylated liposomal doxorubicin.
Influence of dose and animal species on accelerated blood clearance of PEGylated liposomal doxorubicin.
International journal of pharmaceutics (2014-10-05)
Takuya Suzuki, Masako Ichihara, Kenji Hyodo, Eiichi Yamamoto, Tatsuhiro Ishida, Hiroshi Kiwada, Hiroshi Kikuchi, Hiroshi Ishihara
ABSTRACT
We recently demonstrated that Doxil loses its long-circulating properties when injected repeatedly at doses below 2 mg/m(2) in dogs. In studies using other animal species, PEGylated liposomal doxorubicin has been reported not to induce the accelerated blood clearance (ABC) phenomenon. We investigated the issue of whether Doxil can elicit the ABC phenomenon in several species. In minipigs, the ABC phenomenon was induced at 2 mg/m(2). In other animal species, the ABC phenomenon was not observed at higher doses (>2 mg/m(2)), but was observed at much lower doses (0.2 mg/m(2)). The pharmacokinetic profile of a second dose of Doxil reflected the circulating anti-PEG IgM level induced by the first dose. The ABC phenomenon was not observed at the clinically recommended DXR dose (20 mg/m(2)) in any animal species. These results indicate that Doxil can cause the ABC phenomenon in all animals tested, the extent of induction was dependent on the first dose of Doxil, and a higher Doxil dose lessened the ABC phenomenon. The current study results suggest that a careful study design including selection of animal species is important for preclinical studies using PEGylated liposomal formulations even if they contain anticancer drugs that suppress the host immune response.
Product Description
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Sigma-Aldrich
Sulfuric acid solution, 5 mM
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
α-D-Glucose, anhydrous, 96%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
Glucose solution, BioUltra, Molecular Biology, ~20% in H2O
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
D-(+)-Glucose, SAJ special grade, ≥98.0%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications