MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Detecting metabolites of different transition metal-lithospermate B complexes after intravenous injection in rats.
Detecting metabolites of different transition metal-lithospermate B complexes after intravenous injection in rats.
Acta pharmacologica Sinica (2014-07-06)
Ying-Jie Chen, Tse-Yu Chung, Wen-Ying Chen, Chung-Yu Chen, Maw-Rong Lee, Tzyy-Rong Jinn, Jason Tc Tzen
ABSTRACT
Lithospermate B (LSB) isolated from the traditional Chinese medicine danshen (Salvia miltiorrhiza) is an effective Na(+)/K(+)-ATPase inhibitor and used to treat congestive heart failure. The inhibition of LSB on Na(+)/K(+)-ATPase is potentiated by forming complexes with transition metal ions. Here we investigated the safety and metabolites of different transition metal-LSB complexes in rats. LSB complexed with six different transition metal ions (Mg(2+), Zn(2+), Cr(3+), Co(2+), Ni(2+) and Mn(2+)) were prepared. Adult male SD rats were injected with the different metal-LSB complexes (50 mg/kg, iv), and their bile and blood samples were collected. The metabolites of the metal-LSB complexes in the samples were analyzed using mass spectroscopy. In rats injected with LSB complexed with Mg(2+), Zn(2+), Cr(3+), Ni(2+) or Mn(2+), LSB and its four putative metabolites were equivalently detected in their bile samples. Mn(2+)-LSB exhibited distinct metabolite profiles compared with the other four metal-LSB complexes. The four putative metabolites were identified as 3-monomethyl-LSB, 3,3''-dimethyl-LSB, 3,3'''-dimethyl-LSB and 3,3'',3'''-trimethyl-LSB. The tracking of successive bile samples of rats injected with Mg(2+)-LSB, Zn(2+)-LSB and Mn(2+)-LSB concurrently demonstrated that LSB was firstly methylated at position 3, then at position 3'', and, finally, the 3''' hydroxyl group. All rats injected with Co(2+)-LSB died. Zn(2+)-LSB, Cr(3+)-LSB, Ni(2+)-LSB or Mn(2+)-LSB produces identical four methylated metabolites of LSB in rats, and seemed to be as safe as LSB or Mg(2+)-LSB.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sigma-Aldrich
Acetic acid solution, 1 N, 1 M
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium hydroxide solution, 0.2 M
Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC