MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Plasma polyunsaturated fatty acids and periodontal recovery in Taiwanese with periodontitis: a significant relationship.
Plasma polyunsaturated fatty acids and periodontal recovery in Taiwanese with periodontitis: a significant relationship.
Archives of oral biology (2014-05-27)
Sheng-Hung Wang, Hsin-Chia Hung, Chi-Cheng Tsai, Meng-Chuan Huang, Kun-Yen Ho, Yi-Min Wu, Yen-Yun Wang, Ying-Chu Lin
ABSTRACT
Plasma levels of polyunsaturated fatty acids (PUFAs) are different before and after periodontal treatment. Asians and Westerners have significantly different baseline levels of plasma PUFAs. However, no Asian study has reported the effects of nonsurgical treatment on the correlation between periodontal condition and plasma levels of PUFAs. We analyzed whether recovery from periodontitis was correlated with the elevation of plasma fatty acids 3 months after the nonsurgical intervention and with no recommended supplements. Thirty-five Taiwanese patients with periodontitis were recruited. Probing pocket depths (PPDs) and clinical attachment levels (CALs) were measured at baseline and 3 months after the nonsurgical treatment. Plasma levels of fatty acids were determined using gas chromatography. Differences and correlations between plasma fatty acid composition and periodontitis severity at baseline and 3 months after treatment were determined. Twenty-six patients completed the study. At the baseline, PPDs were negatively correlated with plasma n-3 PUFAs (r=-0.52, p<0.01), but at 3 months post intervention, periodontitis severity had declined and the weight percentages of n-3 PUFAs, DPA, and DHA were significantly (p=0.019, 0.005, and 0.037, respectively) higher. The recovery percentages of CALs were positively and significantly correlated with plasma ΔPUFAs and the percentage of Δn-3 PUFAs in ΔPUFAs (r=0.42 and 0.45, respectively; p<0.05 for both). We conclude that a higher weight percentage of n-3 PUFAs in total PUFAs was related to the recovery of CALs 3 months after the nonsurgical periodontal treatment. However, no such relationship was found for PPDs.
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph. Eur.
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichloromethane, analytical standard
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Dichloromethane, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dichloromethane, ≥99.9%
Sigma-Aldrich
Dichloromethane, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dichloromethane, JIS special grade, ≥99.0%
USP
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG