MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effects of anaerobic training on paraoxonase-1 enzyme (PON1) activities of high density lipoprotein subgroups and its relationship with PON1-Q192R phenotype.
Effects of anaerobic training on paraoxonase-1 enzyme (PON1) activities of high density lipoprotein subgroups and its relationship with PON1-Q192R phenotype.
Journal of atherosclerosis and thrombosis (2015-03-05)
Faruk Turgay, Ali Rıza Şişman, Aylin Çeçen Aksu
ABSTRACT
Paraoxonase-1 (PON1) is an antiatherosclerotic enzyme located on high-density lipoprotein (HDL). The effects of anaerobic exercise on PON1 activity are unknown. Here we investigated the effects of anaerobic judo training on three different activities of same PON1 enzyme (TDPON1), including basal PON1, salt-stimulated PON1 (SPON1), and arylesterase (AE) activities, of serum, HDL, and HDL subgroups (HDLs; HDL and its subgroups) and its relationship with PON1-Q192R phenotype (PON1P). Our study included 18 Turkish national female judoists (mean age: 17.9 ± 0.8 years). Before and after 5 months of anaerobic training, critical speed (CS), TDPON1 activities, cholesterol levels in the serum and supernatants of HDLs obtained by polyethylene glycol, and other major blood lipids and lipoproteins (BLLPs) including triglycerides were determined using blood samples taken after overnight fasting. PON1P groups (PGs) were categorized as QQ (QG; persons with low activity) and R carriers (QR + RR) (RG; persons with high activity) according to SPON1/AE activity ratios. The results were considered statistically significant at P ≤ 0.05. Anaerobic training resulted in significantly increased the cholesterol levels of HDLs (except HDL2-C) in all subjects, but not HDLs-C in PGs. Anaerobic training resulted in significant increases in most TDPON1 activities of serum and HDLs in all subjects and (except AE) in PGs, whereas SPON1 and HDL2 AE activities increased only in the RG, which was related to PON1P. However, PON1P was not related to other measured markers, including basal BLLP profiles. Anaerobic training improved most TDPON1 activities of serum and HDLs and HDLs -C levels (except HDL2-C) in all subjects, but not HDLs-C in PGs. The beneficial effects of anaerobic training on SPON1 and HDL2 AE activities were depend on PON1P. The lack of response of HDL2-C to anaerobic exercise will require further research.
Product Description
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Lysine monohydrochloride, SAJ special grade, ≥99.0%
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard