MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Mechanism of reversible self-association of a monoclonal antibody: role of electrostatic and hydrophobic interactions.
Mechanism of reversible self-association of a monoclonal antibody: role of electrostatic and hydrophobic interactions.
Journal of pharmaceutical sciences (2014-11-20)
Reza Esfandiary, Arun Parupudi, Jose Casas-Finet, Dhanesh Gadre, Hasige Sathish
ABSTRACT
Reversible self-association of protein therapeutics, the phenomenon of formation of native reversible oligomeric species as a result of noncovalent intermolecular interactions, can add additional manufacturing, stability, delivery, and safety complexities in biopharmaceutical development. Its early detection, characterization, and mitigation can therefore contribute to the success of drug development. A variety of structural and environmental factors can contribute to the modulation of self-association with mechanisms still elusive in some cases due to the inherent structural complexity of proteins. By combining the capabilities of dynamic and static light scattering techniques, the modulatory effects of a variety of solution conditions on a model IgG1's (mAbA) intermolecular interactions have been utilized to derive mechanism of its self-association at relatively low-protein concentration. The analysis of the effect of pH, buffer type, Hofmeister salts, and aromatic amino acids utilizing light scattering supported a combined role of hydrophobic and electrostatic interactions in mAbA self-association. Fitting of the data into the equilibrium models obtained from the multiangle static light scattering provided the enthalpic and entropic contributions of self-association, highlighting the more dominant effect of electrostatic interactions. In addition, studies of the Fab and Fc fragments of mAbA suggested the key role of the former in observed self-association.
Sigma-Aldrich
Toluene, JIS 1000, suitable for residue analysis, ≥99.8%
Sigma-Aldrich
Toluene, SAJ first grade, ≥99.0%
Supelco
Toluene, HPLC grade, 99.8%
Arginine, European Pharmacopoeia (EP) Reference Standard
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Toluene, analytical standard
Sigma-Aldrich
Toluene, JIS special grade, ≥99.5%
Sigma-Aldrich
Toluene, JIS 300, suitable for residue analysis, ≥99.8%
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material