MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Qualitative and quantitative determination of peptides related to celiac disease in mixtures derived from different methods of simulated gastrointestinal digestion of wheat products.
Qualitative and quantitative determination of peptides related to celiac disease in mixtures derived from different methods of simulated gastrointestinal digestion of wheat products.
Analytical and bioanalytical chemistry (2014-05-21)
Barbara Prandi, Andrea Faccini, Tullia Tedeschi, Alessandro Cammerata, Daniela Sgrulletta, Maria Grazia D'Egidio, Gianni Galaverna, Stefano Sforza
ABSTRACT
During wheat digestion, gluten-derived proteolytic resistant peptides are generated, some of them involved in celiac disease. In vitro digestion models able to mimic the peptides generated in the human gastrointestinal tract are extremely useful to assess the pathogenicity of wheat-derived products. In this paper, samples belonging to three different durum wheat varieties were taken at six different steps of the pasta production chain and two different digestion models present in the literature were assessed on the different samples: a more complex one using artificial fluids simulating the exact composition of digestive juices, and a simplified method based on a peptic-tryptic/chymotryptic treatment of wheat ethanolic extract. An extensive characterization of the peptides generated using two in vitro digestion models was performed through LC-MS/MS techniques and the two methods were compared in order to evaluate qualitative and quantitative differences and their possible implications for varietal screening. Strong differences in the type of peptides produced with the two methods were detected, indicating that the simplified method can still be used for a varietal screening but is not representative of the peptides really generated after physiological human digestion. Results indicate a clear necessity of physiologically accurate models for simulating human gastrointestinal digestion of wheat products.
Sigma-Aldrich
N,N-Diisopropylethylamine, 99.5%, biotech. grade
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
USP
Glucosamine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Glucosamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Glucosamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Calcium chloride solution, 0.025 M
Sigma-Aldrich
Calcium chloride solution, 3.2 mM
Sigma-Aldrich
Potassium thiocyanate solution, 0.1 M
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Supelco
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard