MilliporeSigma
  • Herbal preparation extract for skin after radiotherapy treatment. Part One--Preclinical tests.
Herbal preparation extract for skin after radiotherapy treatment. Part One--Preclinical tests.
Acta poloniae pharmaceutica (2014-11-05)
Agnieszka Skalska-Kamińska, Anna Woźniak, Roman Paduch, Ryszard Kocjan, Robert Rejdak
ABSTRACT
Naran R is a herbal composition made of Plantago lanceolate folium, Malvae arboreae flos, Calendulae flos, Chamomillae inflorescentia, Lamii albi flos to prepare compresses or to wash skin with inflammations. The extract of this preparation is mixed to be applied as an ointment on patients' skin after radiotherapy. Experiments performed in vitro are part of pre-clinical tests with Naran R ointment. This study examined the impact of the plant composition for ethanol-water extract on human skin fibroblasts (HSF) culture. Samples of extract, prepared from patented amounts of herbs, were in the range of 25-225 μg/mL. Six methods were applied: standard spectrophotometric 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) assay, neutral red (NR) uptake assay, DPPH free radical scavenging test, labeling of cytoskeleton F-actin, staining of argyrophilic nucleolar organizer regions (AgNORs) and trypan blue coloration. The extract concentration 75 μg/mL was established as safe for application on human skin. In labeling of F-actin with rhodamine-phalloidin dye at this concentration the cytoskeleton was stable. The extract did not influence the membrane stability and had positive influence on the proliferation activity. It was confirmed in AgNOR test during incubation with extract, which led to formation of larger amount of smaller nucleolins. In DPPH scavenging activity test, the extract revealed over 8% higher free-radical scavenging activity in comparison to control. After trypan blue staining, the extract in concentration 125 μg/mL significantly lowered the cell viability. When the cytotoxic and anti-proliferative activity of the extracts were analyzed, MTT and Neutral Red (NR) methods were used. The cells' viability was maintained on a constant level (80-110%) after 24, 48 and 72 h of incubation. During all time of NR test (72 h) and even when 225 μg/mL of extract was applied, the viability of cells was in range 80-110% of control. Positive influence of the extract on investigated cells structure and proliferation, lack of toxicity and increasing anti-oxidant activity enable to consider this preparation as a natural remedy with potential application in skin therapy after radiation.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard