MilliporeSigma
  • Design, Synthesis, and Photophysical Properties of Pyrroloquinoline-Based Compounds Showing Strong Blue Fluorescence as Potential Dyes for Biomedical Applications.
Design, Synthesis, and Photophysical Properties of Pyrroloquinoline-Based Compounds Showing Strong Blue Fluorescence as Potential Dyes for Biomedical Applications.
ChemMedChem (2015-10-09)
Davide Carta, Anna Balasso, Paolo Caliceti, Maria Grazia Ferlin
ABSTRACT
A small library of 3-ethylpyrrolo[3,2-f]quinoline derivatives was synthesized to identify a novel class of dyes for use in biological studies. According to the spectroscopic analyses performed to evaluate the fluorimetric parameters of quantum yield and brightness, 7-methyl- and 6,7-dimethylpyrroloquinolin(9)one derivatives were found to be the best blue luminescent dyes for biological applications. To enhance the luminescence profiles and to obtain probes that could be conjugated to functional groups of supramolecular drug delivery systems, these compounds were further modified at position 3 to obtain 3-heptanoic acid and 3-aminohexylpyrroloquinolin(9)one methylated derivatives. The most brilliant 6,7-dimethyl-3-aminohexylpyrroloquinolinone hydrochloride was conjugated to pullulan, a biocompatible polysaccharide used to produce colloidal systems for drug delivery. Comparative studies showed that this compound can be properly exploited as a blue fluorescent label in biological investigations, namely cell trafficking and pharmacokinetics/biodistribution studies. These molecules possess higher fluorescence efficiency than commercial dyes in biological media, making them suitable alternatives to commercially available products in current use.
Sigma-Aldrich
D-Mannitol, SAJ first grade, ≥99.0%
Sigma-Aldrich
D-Mannitol, SAJ special grade, ≥99.0%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Sodium (meta)periodate, ≥99.0%
Sigma-Aldrich
Sodium (meta)periodate, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Sodium cyanoborohydride solution, 1.0 M in THF
Sigma-Aldrich
Sodium cyanoborohydride, reagent grade, 95%
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Tetramethylsilane, NMR grade, ACS reagent, ≥99.9%
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
Water, BioPerformance Certified