MilliporeSigma
  • Spin Exchange Interaction in Substituted Copper Phthalocyanine Crystalline Thin Films.
Spin Exchange Interaction in Substituted Copper Phthalocyanine Crystalline Thin Films.
Scientific reports (2015-11-13)
Naveen Rawat, Zhenwen Pan, Cody J Lamarche, Anthony Wetherby, Rory Waterman, Takahisa Tokumoto, Judy G Cherian, Randall L Headrick, Stephen A McGill, Madalina I Furis
ABSTRACT
The origins of spin exchange in crystalline thin films of Copper Octabutoxy Phthalocyanine (Cu-OBPc) are investigated using Magnetic Circular Dichroism (MCD) spectroscopy. These studies are made possible by a solution deposition technique which produces highly ordered films with macroscopic grain sizes suitable for optical studies. For temperatures lower than 2 K, the contribution of a specific state in the valence band manifold originating from the hybridized lone pair in nitrogen orbitals of the Phthalocyanine ring, bears the Brillouin-like signature of an exchange interaction with the localized d-shell Cu spins. A comprehensive MCD spectral analysis coupled with a molecular field model of a σπ - d exchange analogous to sp-d interactions in Diluted Magnetic Semiconductors (DMS) renders an enhanced Zeeman splitting and a modified g-factor of -4 for the electrons that mediate the interaction. These studies define an experimental tool for identifying electronic states involved in spin-dependent exchange interactions in organic materials.
Sigma-Aldrich
Copper(II) 1,4,8,11,15,18,22,25-octabutoxy-29H,31H-phthalocyanine, Dye content 95 %
Pricing and availability is not currently available.