MilliporeSigma
  • Bactericidal Compounds Controlling Growth of the Plant Pathogen Pseudomonas syringae pv. actinidiae, Which Forms Biofilms Composed of a Novel Exopolysaccharide.
Bactericidal Compounds Controlling Growth of the Plant Pathogen Pseudomonas syringae pv. actinidiae, Which Forms Biofilms Composed of a Novel Exopolysaccharide.
Applied and environmental microbiology (2015-04-05)
Shirin Ghods, Ian M Sims, M Fata Moradali, Bernd H A Rehm
ABSTRACT
Pseudomonas syringae pv. actinidiae is the major cause of bacterial canker and is a severe threat to kiwifruit production worldwide. Many aspects of the disease caused by P. syringae pv. actinidiae, such as the pathogenicity-relevant formation of a biofilm composed of extracellular polymeric substances (EPSs), are still unknown. Here, a highly virulent strain of P. syringae pv. actinidiae, NZ V-13, was studied with respect to biofilm formation and architecture using a flow cell system combined with confocal laser scanning microscopy. The biofilm formed by P. syringae pv. actinidiae NZ V-13 was heterogeneous, consisting of a thin cellular base layer 5 μm thick and microcolonies with irregular structures. The major component of the EPSs produced by P. syringae pv. actinidiae NZ V-13 bacteria was isolated and identified to be an exopolysaccharide. Extensive compositional and structural analysis showed that rhamnose, fucose, and glucose were the major constituents, present at a ratio of 5:1.5:2. Experimental evidence that P. syringae pv. actinidiae NZ V-13 produces two polysaccharides, a branched α-d-rhamnan with side chains of terminal α-d-Fucf and an α-d-1,4-linked glucan, was obtained. The susceptibility of the cells in biofilms to kasugamycin and chlorine dioxide was assessed. About 64 and 73% of P. syringae pv. actinidiae NZ V-13 cells in biofilms were killed when kasugamycin and chlorine dioxide were used at 5 and 10 ppm, respectively. Kasugamycin inhibited the attachment of P. syringae pv. actinidiae NZ V-13 to solid surfaces at concentrations of 80 and 100 ppm. Kasugamycin was bacteriostatic against P. syringae pv. actinidiae NZ V-13 growth in the planktonic mode, with the MIC being 40 to 60 ppm and a bactericidal effect being found at 100 ppm. Here we studied the formation, architecture, and composition of P. syringae pv. actinidiae biofilms as well as used the biofilm as a model to assess the efficacies of bactericidal compounds.
Sigma-Aldrich
Sulfuric acid solution, 0.5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Diethyl azodicarboxylate solution, 40 wt. % in toluene
Pricing and availability is not currently available.
Sigma-Aldrich
Anthrone, ACS reagent, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid, 99.999%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 0.25 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 1.5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 0.01 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid, JIS special grade, ≥95.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 0.1 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 0.05 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 5 mM
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid solution, 0.025 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfuric acid, SAJ first grade, ≥95.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Deuterium oxide, 99 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Pricing and availability is not currently available.
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Pricing and availability is not currently available.
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Pricing and availability is not currently available.