MilliporeSigma
  • Homozygous mutations in MFN2 cause multiple symmetric lipomatosis associated with neuropathy.
Homozygous mutations in MFN2 cause multiple symmetric lipomatosis associated with neuropathy.
Human molecular genetics (2015-06-19)
Sarah L Sawyer, Andy Cheuk-Him Ng, A Micheil Innes, Justin D Wagner, David A Dyment, Martine Tetreault, Jacek Majewski, Kym M Boycott, Robert A Screaton, Garth Nicholson
ABSTRACT
Multiple symmetric lipomatosis (MSL) is a mitochondrial disorder with impaired brown fat metabolism that has been associated with MERRF mutations in some, but not all, patients. We studied a sibling pair and an unrelated indiviadual who presented with MSL and neuropathy to determine the genetic etiology of this disorder in patients who did not carry the MSL-associated MERRF mutation. Whole-exome sequencing was performed on the siblings, and a rare, shared homozygous mutation in MFN2 (c.2119C>T: p.R707W) was identified. The mutation was not present in their healthy siblings. In silico programs predict it to be pathogenic, and heterozygous carriers of the MFN2 p.R707W substitution are known to have Charcot-Marie-Tooth (CMT) disease. A third, unrelated patient with multiple symmetrical lipomatosis and neuropathy also harbored the same homozygous mutation and had been previously diagnosed with CMT. Functional studies in patient fibroblasts demonstrate that the p.R707W substitution impairs homotypic (MFN2-MFN2) protein interactions required for normal activity and renders mitochondria prone to perinuclear aggregation. These findings show that homozygous mutations at p.R707W in MFN2 are a novel cause of multiple symmetrical lipomatosis.
Sigma-Aldrich
MISSION® esiRNA, targeting human MFN1
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mfn1
Pricing and availability is not currently available.