MilliporeSigma
4-ketoproline: An electrophilic proline analog for bioconjugation.
Biopolymers (2015-02-07)
Amit Choudhary, Kimberli J Kamer, Matthew D Shoulders, Ronald T Raines
ABSTRACT
Installing an electrophilic amino-acid residue can engender a peptide or protein with chemoselective reactivity. Such a modification to collagen, which is the most abundant protein in animals, could facilitate the development of new biomaterials. Collagen has an abundance of proline-like residues. Here, we report on the incorporation of an electrophilic proline congener, (2S)-4-ketoproline (Kep), into a collagen-mimetic peptide (CMP). An ab initio conformational analysis of Kep revealed its potential to be accommodated within a collagen triple helix. A synthetic CMP containing a Kep residue was indeed able to form a stable triple helix. Moreover, the condensation of its carbonyl group with aminooxy-biotin did not compromise the conformational stability of the triple helix. These data encourage the use of 4-ketoproline as an electrophilic congener of proline.
Sigma-Aldrich
Cytidine 5′-monophosphate, Sigma Grade, ≥99% (HPLC), synthetic, powder
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Pricing and availability is not currently available.
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Pricing and availability is not currently available.