MilliporeSigma
  • Understanding the structure and function of catalases: clues from molecular evolution and in vitro mutagenesis.
Understanding the structure and function of catalases: clues from molecular evolution and in vitro mutagenesis.
Progress in biophysics and molecular biology (1999-08-14)
M Zámocký, F Koller
ABSTRACT
This review gives an overview about the structural organisation of different evolutionary lines of all enzymes capable of efficient dismutation of hydrogen peroxide. Major potential applications in biotechnology and clinical medicine justify further investigations. According to structural and functional similarities catalases can be divided in three subgroups. Typical catalases are homotetrameric haem proteins. The three-dimensional structure of six representatives has been resolved to atomic resolution. The central core of each subunit reveals a characteristic "catalase fold", extremely well conserved among this group. In the native tetramer structure pairs of subunits tightly interact via exchange of their N-terminal arms. This pseudo-knot structures implies a highly ordered assembly pathway. A minor subgroup ("large catalases") possesses an extra flavodoxin-like C-terminal domain. A > or = 25 A long channel leads from the enzyme surface to the deeply buried active site. It enables rapid and selective diffusion of the substrates to the active center. In several catalases NADPH is tightly bound close to the surface. This cofactor may prevent and reverse the formation of compound II, an inactive reaction intermediate. Bifunctional catalase-peroxidase are haem proteins which probably arose via gene duplication of an ancestral peroxidase gene. No detailed structural information is currently available. Even less is know about manganese catalases. Their di-manganese reaction centers may be evolutionary.
Sigma-Aldrich
Catalase Assay Kit, sufficient for ≥100 tests enzymatic, determination of catalase activity in tissues and cells
Pricing and availability is not currently available.