MilliporeSigma
  • HucMSC-Exosome Mediated-Wnt4 Signaling Is Required for Cutaneous Wound Healing.
HucMSC-Exosome Mediated-Wnt4 Signaling Is Required for Cutaneous Wound Healing.
Stem cells (Dayton, Ohio) (2014-06-26)
Bin Zhang, Mei Wang, Aihua Gong, Xu Zhang, Xiaodan Wu, Yanhua Zhu, Hui Shi, Lijun Wu, Wei Zhu, Hui Qian, Wenrong Xu
ABSTRACT
Mesenchymal stem cell-derived exosomes (MSC-Ex) play important roles in tissue injury repair, however, the roles of MSC-Ex in skin damage repair and its mechanisms are largely unknown. Herein, we examined the benefit of human umbilical cord MSC-derived exosome (hucMSC-Ex) in cutaneous wound healing using a rat skin burn model. We found that hucMSC-Ex-treated wounds exhibited significantly accelerated re-epithelialization, with increased expression of CK19, PCNA, collagen I (compared to collagen III) in vivo. HucMSC-Ex promoted proliferation and inhibited apoptosis of skin cells after heat-stress in vitro. We also discovered that Wnt4 was contained in hucMSC-Ex, and hucMSC-Ex-derived Wnt4 promoted β-catenin nuclear translocation and activity to enhance proliferation and migration of skin cells, which could be reversed by β-catenin inhibitor ICG001. In vivo studies confirmed that the activation of Wnt/β-catenin by hucMSC-Ex played a key role in wound re-epithelialization and cell proliferation. Furthermore, knockdown of Wnt4 in hucMSC-Ex abrogated β-catenin activation and skin cell proliferation and migration in vitro. The in vivo therapeutic effects were also inhibited when the expression of Wnt4 in hucMSC-Ex was interfered. In addition, the activation of AKT pathway by hucMSC-Ex was associated with the reduction of heat stress-induced apoptosis in rat skin burn model. Collectively, our findings indicate that exosome-delivered Wnt4 provides new aspects for the therapeutic strategy of MSCs in cutaneous wound healing. Stem Cells 2015;33:2158-2168.
Sigma-Aldrich
MISSION® esiRNA, targeting human WNT4
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Wnt4
Pricing and availability is not currently available.