MilliporeSigma
  • Functional evaluation of gene silencing on macrophages derived from U937 cells using interference RNA (shRNA) in a model of macrophages infected with Leishmania (Viannia) braziliensis.
Functional evaluation of gene silencing on macrophages derived from U937 cells using interference RNA (shRNA) in a model of macrophages infected with Leishmania (Viannia) braziliensis.
Parasitology (2015-10-08)
Clemencia Ovalle-Bracho, Diana A Londoño-Barbosa, Carlos Franco-Muñoz, Carlos Clavijo-Ramírez
ABSTRACT
Leishmaniasis development is multifactorial; nonetheless, the establishment of the infection, which occurs by the survival and replication of the parasite inside its main host cell, the macrophage, is mandatory. Thus, the importance of studying the molecular mechanisms involved in the Leishmania-macrophage interaction is highlighted. The aim of this study was to characterize a cellular model of macrophages derived from U937 cells that would allow for the identification of infection phenotypes induced by genetic silencing with interference RNA in the context of macrophages infected with Leishmania (Viannia) braziliensis. The model was standardized by silencing an exogenous gene (gfp), an endogenous gene (lmna) and a differentially expressed gene between infected and non-infected macrophages (gro-β). The silencing process was successful for the three genes studied, obtaining reductions of 88·9% in the GFP levels, 87·5% in LMNA levels and 74·4% for Gro-β with respect to the corresponding control cell lines. The cell model revealed changes in the infection phenotype of the macrophages in terms of number of amastigotes per infected macrophage, number of amastigotes per sampled macrophage and percentage of infected macrophages as a result of gene silencing. Thus, this cell model constitutes a research platform for the study of parasite-host interactions and for the identification of potentially therapeutic targets.
Sigma-Aldrich
Sodium butyrate, Vetec, reagent grade, 99%
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium butyrate, 98%
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)