MilliporeSigma
Antimicrobial activity of synthetic bactenecin.
Biotechnology therapeutics (1989-01-01)
B Gallis, J Mehl, K S Prickett, J A Martin, J Merriam, C J March, D P Cerretti
ABSTRACT
Bactenecin is an antimicrobial peptide isolated from bovine neutrophils. Bactenecin was synthesized by solid-phase peptide synthesis and renatured to a fully disulfide bonded form. The peptide inhibits the growth of Escherichia coli and Staphylococcus aureus at the same concentration reported for the peptide purified from bovine neutrophils. Bactenecin inhibits the growth of other medically important bacteria and yeast, and it kills the fungus Trichophyton rubrum. Acetylation and amidation of the amino- and carboxy-termini of bactenecin do not change its potency, while replacement of its two cysteine residues with serine decreases the potency.
Sigma-Aldrich
Bactenecin, ≥95% (HPLC)
Pricing and availability is not currently available.