MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Effect of bore fluid composition on microstructure and performance of a microporous hollow fibre membrane as a cation-exchange substrate.
Effect of bore fluid composition on microstructure and performance of a microporous hollow fibre membrane as a cation-exchange substrate.
Journal of chromatography. A (2015-04-05)
R A Lazar, I Mandal, N K H Slater
ABSTRACT
Micro-capillary film (MCF) membranes are effective platforms for bioseparations and viable alternatives to established packed bed and membrane substrates at the analytical and preparative chromatography scales. Single hollow fibre (HF) MCF membranes with varied microstructures were produced in order to evaluate the effect of the bore fluid composition used during hollow fibre extrusion on their structure and performance as cation-exchange adsorbers. Hollow fibres were fabricated from ethylene-vinyl alcohol (EVOH) copolymer through solution extrusion followed by nonsolvent induced phase separation (NIPS) using bore fluids of differing composition (100wt.% N-methyl-2-pyrrolidone (NMP), 100wt.% glycerol, 100wt.% water). All HFs displayed highly microporous and mesoporous microstructures, with distinct regions of pore size <1μm, 5-15μm and up to 50μm in diameter, depending upon proximity to the bore fluid. Scanning electron microscopy (SEM) revealed skins of pore size <1μm at the inner surface of HFs produced with water and glycerol, while NMP bore fluid resulted in a skinless inner HF surface. The HFs were modified for chromatography by functionalising the polymer surface hydroxyl groups with sulphonic acid (SP) groups to produce cation-exchange adsorbers. The maximum binding capacities of the HFs were determined by frontal analysis using lysozyme solutions (0.05-100mgml(-1)) for a flow rate of 1.0mlmin(-1). The NMP-HF-SP module displayed the largest maximum lysozyme binding capacity of all the fibres produced (40.3mg lysozyme/ml adsorbent volume), a nearly 2-fold increase over the glycerol and 10-fold increase over the water variants at the same sample flow rate. The importance of NMP as a bore fluid to hollow fibre membrane performance as a result of inner surface porosity was established with a view to applying this parameter for the optimisation of multi-capillary MCF performance in future studies.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
3-Amino-1-propanesulfonic acid, 97%
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Cyanuric chloride, 99%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, SAJ first grade, ≥98.0%
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
SAFC
Tromethamine
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ≥99.8%
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, JIS special grade, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ≥99.8%
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sigma-Aldrich
Sodium hydroxide solution, 0.2 M
Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%