MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Impedance technology reveals correlations between cytotoxicity and lipophilicity of mono and bimetallic phosphine complexes.
Impedance technology reveals correlations between cytotoxicity and lipophilicity of mono and bimetallic phosphine complexes.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine (2015-04-02)
P Fonteh, A Elkhadir, B Omondi, I Guzei, J Darkwa, D Meyer
ABSTRACT
Label free impedance technology enables the monitoring of cell response patterns post treatment with drugs or other chemicals. Using this technology, a correlation between the lipophilicity of metal complexes and the degree of cytotoxicity was observed. Au(L1)Cl (1), AuPd(L1)(SC4H8)Cl3 (1a) and Au(L2)Cl (2) [L1 = diphenylphosphino-2-pyridine; L2 = 2-(2-(diphenylphosphino)ethyl)-pyridine] were synthesised, in silico drug-likeness and structure-activity relationship monitored using impedance technology. Dose dependent changes in cytotoxicity were observed for the metal complexes resulting in IC50s of 12.5 ± 2.5, 18.3 ± 8.3 and 16.9 ± 0.5 µM for 1, 1a and 2 respectively in an endpoint assay. When a real time impedance assay was used, dose-dependent responses depicting patterns that suggested slower uptake (at a toxic 20 µM) and faster recovery of the cells (at the less toxic 10 µM) of the bimetallic complex (1a) compared to the monometallic complexes (1 and 2) was observed. These data agreed with the ADMET findings of lower aqueous solubility of 1a and non-ideal lipophilicity (AlogP98 of 6.55) over more water soluble 1 and 2 with ideal lipophilicity (4.91 and 5.03 respectively) values. The additional coordination of a Pd atom to the nitrogen atom of a pyridine ring, the sulfur atom of a tetrahydrothiophene moiety and two chlorine atoms in 1a could be contributing to the observed differences when compared to the monometallic complexes. This report presents impedance technology as a means of correlating drug-likeness of lipophilic phosphine complexes containing similar backbone structures and could prove valuable in filtering drug-like compounds in a drug discovery project.
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C