MilliporeSigma
  • Highly efficient CRISPR/Cas9-mediated transgene knockin at the H11 locus in pigs.
Highly efficient CRISPR/Cas9-mediated transgene knockin at the H11 locus in pigs.
Scientific reports (2015-09-19)
Jinxue Ruan, Hegang Li, Kui Xu, Tianwen Wu, Jingliang Wei, Rong Zhou, Zhiguo Liu, Yulian Mu, Shulin Yang, Hongsheng Ouyang, Ruby Yanru Chen-Tsai, Kui Li
ABSTRACT
Transgenic pigs play an important role in producing higher quality food in agriculture and improving human health when used as animal models for various human diseases in biomedicine. Production of transgenic pigs, however, is a lengthy and inefficient process that hinders research using pig models. Recent applications of the CRISPR/Cas9 system for generating site-specific gene knockout/knockin models, including a knockout pig model, have significantly accelerated the animal model field. However, a knockin pig model containing a site-specific transgene insertion that can be passed on to its offspring remains lacking. Here, we describe for the first time the generation of a site-specific knockin pig model using a combination of CRISPR/Cas9 and somatic cell nuclear transfer. We also report a new genomic "safe harbor" locus, named pH11, which enables stable and robust transgene expression. Our results indicate that our CRISPR/Cas9 knockin system allows highly efficient gene insertion at the pH11 locus of up to 54% using drug selection and 6% without drug selection. We successfully inserted a gene fragment larger than 9 kb at the pH11 locus using the CRISPR/Cas9 system. Our data also confirm that the gene inserted into the pH11 locus is highly expressed in cells, embryos and animals.
Product Description
Roche
DIG-High Prime DNA Labeling and Detection Starter Kit II, sufficient for 12 labeling reactions (10 ng to 3 μg per assay), sufficient for 24 blots (blots of 100 cm2)
Pricing and availability is not currently available.