MilliporeSigma
  • P21-activated kinase 7 mediates cisplatin-resistance of esophageal squamous carcinoma cells with Aurora-A overexpression.
P21-activated kinase 7 mediates cisplatin-resistance of esophageal squamous carcinoma cells with Aurora-A overexpression.
PloS one (2014-12-02)
Shun He, Min Feng, Mei Liu, Shangbin Yang, Shuang Yan, Wei Zhang, Zaozao Wang, Chenfei Hu, Qing Xu, Lechuang Chen, Hongxia Zhu, Ningzhi Xu
ABSTRACT
Aurora-A overexpression is common in various types of cancers and has been shown to be involved in tumorigenesis through different signaling pathways, yet how the deregulation affects cancer therapeutics remains elusive. Here we showed that overexpression of Aurora-A rendered esophageal cancer cells resistance to cisplatin (CDDP) by inhibiting apoptosis. By using an apoptosis array, we identified a downstream gene, p21-activated kinase 7 (PAK7). PAK7 was upregulated by Aurora-A overexpression at both mRNA and protein levels. Importantly, the expression levels of Aurora-A and PAK7 were correlated in ESCC primary samples. Chromatin immunoprecipitation (ChIP) assay revealed that binding of E2F1 to the promoter of PAK7 was significantly enhanced upon Aurora-A activation, and knockdown of transcription factor E2F1 decreased PAK7 expression, suggesting that Aurora-A regulated PAK7 through E2F1. Furthermore, we demonstrated that PAK7 knockdown led to increased apoptosis, and Aurora-A-induced resistance to CDDP was reversed by downregulation of PAK7, suggesting PAK7 was a downstream player of Aurora-A that mediated chemoresistance of ESCC cells to CDDP. Our data suggest that PAK7 may serve as an attractive candidate for therapeutics in ESCC patients with Aurora-A abnormality.
Sigma-Aldrich
PAK7, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Pricing and availability is not currently available.