MilliporeSigma
  • Bisphenol A bis(2,3-dihydroxypropyl) ether (BADGE.2H2O) induces orphan nuclear receptor Nur77 gene expression and increases steroidogenesis in mouse testicular Leydig cells.
Bisphenol A bis(2,3-dihydroxypropyl) ether (BADGE.2H2O) induces orphan nuclear receptor Nur77 gene expression and increases steroidogenesis in mouse testicular Leydig cells.
Molecules and cells (2008-06-06)
Seung-Won Ahn, Balachandar Nedumaran, Yuanbin Xie, Don-Kyu Kim, Yong Deuk Kim, Hueng-Sik Choi
ABSTRACT
Bisphenol A bis (2,3-dihydroxypropyl) ether (BADGE.2H(2)O) is a component of commercial liquid epoxy resins commonly used in the food-packing industry and in dental sealants. There is evidence that it has significant estrogenic activity. Nur77 plays a crucial role in the regulation of certain genes involved in LH-mediated steroidogenesis in testicular Leydig cells. It was previously demonstrated that Bisphenol A (BPA) stimulates Nur77 gene induction and steroidogenesis. In this study, we investigated the effects of BADGE.2H(2)O on Nur77 gene expression and steroidogenesis. Northern blot analysis showed that it increased the expression of Nur77 mRNA and protein, and transient transfection assays demonstrated that it increased the promoter activity and transactivation of Nur77. It also increased the expression of certain steroidogenic genes, such as StAR and 3 beta-HSD. Finally, over-expression of a dominant negative Nur77 cDNA via adenoviral infection reduced BADGE.2H(2)O-mediated progesterone biosynthesis. These results indicate that BADGE.2H(2)O disrupts testicular steroidogenesis by increasing Nur77 gene expression.
Product Description
Supelco
Bisphenol A bis(2,3-dihydroxypropyl) ether, analytical standard
Pricing and availability is not currently available.