MilliporeSigma
  • The Histone Demethylase PHF8 Is Essential for Endothelial Cell Migration.
The Histone Demethylase PHF8 Is Essential for Endothelial Cell Migration.
PloS one (2016-01-12)
Lunda Gu, Juliane Hitzel, Franziska Moll, Christoph Kruse, Randa Abdel Malik, Jens Preussner, Mario Looso, Matthias S Leisegang, Dieter Steinhilber, Ralf P Brandes, Christian Fork
ABSTRACT
Epigenetic marks critically control gene expression and thus the cellular activity state. The functions of many epigenetic modifiers in the vascular system have not yet been studied. We screened for histone modifiers in endothelial cells and observed a fairly high expression of the histone plant homeodomain finger protein 8 (PHF8). Given its high expression, we hypothesize that this histone demethylase is important for endothelial cell function. Overexpression of PHF8 catalyzed the removal of methyl-groups from histone 3 lysine 9 (H3K9) and H4K20, whereas knockdown of the enzyme increased H3K9 methylation. Knockdown of PHF8 by RNAi also attenuated endothelial proliferation and survival. As a functional readout endothelial migration and tube formation was studied. PHF8 siRNA attenuated the capacity for migration and developing of capillary-like structures. Given the impact of PHF8 on cell cycle genes, endothelial E2F transcription factors were screened, which led to the identification of the gene repressor E2F4 to be controlled by PHF8. Importantly, PHF8 maintains E2F4 but not E2F1 expression in endothelial cells. Consistently, chromatin immunoprecipitation revealed that PHF8 reduces the H3K9me2 level at the E2F4 transcriptional start site, demonstrating a direct function of PHF8 in endothelial E2F4 gene regulation. Conclusion: PHF8 by controlling E2F4 expression maintains endothelial function.
Sigma-Aldrich
Oligo(dT)23, Anchored, 70 μM in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Pricing and availability is not currently available.