MilliporeSigma
  • The experimental power of FR900359 to study Gq-regulated biological processes.
The experimental power of FR900359 to study Gq-regulated biological processes.
Nature communications (2015-12-15)
Ramona Schrage, Anna-Lena Schmitz, Evelyn Gaffal, Suvi Annala, Stefan Kehraus, Daniela Wenzel, Katrin M Büllesbach, Tobias Bald, Asuka Inoue, Yuji Shinjo, Ségolène Galandrin, Naveen Shridhar, Michael Hesse, Manuel Grundmann, Nicole Merten, Thomas H Charpentier, Matthew Martz, Adrian J Butcher, Tanja Slodczyk, Sylvain Armando, Maike Effern, Yoon Namkung, Laura Jenkins, Velten Horn, Anne Stößel, Harald Dargatz, Daniel Tietze, Diana Imhof, Céline Galés, Christel Drewke, Christa E Müller, Michael Hölzel, Graeme Milligan, Andrew B Tobin, Jesús Gomeza, Henrik G Dohlman, John Sondek, T Kendall Harden, Michel Bouvier, Stéphane A Laporte, Junken Aoki, Bernd K Fleischmann, Klaus Mohr, Gabriele M König, Thomas Tüting, Evi Kostenis
ABSTRACT
Despite the discovery of heterotrimeric αβγ G proteins ∼25 years ago, their selective perturbation by cell-permeable inhibitors remains a fundamental challenge. Here we report that the plant-derived depsipeptide FR900359 (FR) is ideally suited to this task. Using a multifaceted approach we systematically characterize FR as a selective inhibitor of Gq/11/14 over all other mammalian Gα isoforms and elaborate its molecular mechanism of action. We also use FR to investigate whether inhibition of Gq proteins is an effective post-receptor strategy to target oncogenic signalling, using melanoma as a model system. FR suppresses many of the hallmark features that are central to the malignancy of melanoma cells, thereby providing new opportunities for therapeutic intervention. Just as pertussis toxin is used extensively to probe and inhibit the signalling of Gi/o proteins, we anticipate that FR will at least be its equivalent for investigating the biological relevance of Gq.
Sigma-Aldrich
9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F, solution, 10 mg/mL in methyl acetate
Pricing and availability is not currently available.