MilliporeSigma
  • Evaluation of garlic cultivars for polyphenolic content and antioxidant properties.
Evaluation of garlic cultivars for polyphenolic content and antioxidant properties.
PloS one (2013-11-16)
Shuxia Chen, Xiaoqing Shen, Siqiong Cheng, Panpan Li, Junna Du, Yanxia Chang, Huanwen Meng
ABSTRACT
Two phenolic compound parameters (total phenolic and flavonoid contents) and 5 antioxidant parameters (DPPH [2, 2-diphenyl-1-picrylhydrazyl] radical scavenging activity, HRSC (hydroxyl radical scavenging capacity), FRAP (ferric ion reducing antioxidant power), CUPRAC (cupric ion reducing antioxidant capacity), and MCA (metal chelating activity) were measured in bulbs and bolts of 43 garlic cultivars. The bulbs of cultivar '74-x' had the highest phenolic content (total phenolic, flavonoids) and the strongest antioxidant capacity (DPPH, FRAP, and CUPRAC), followed by bulbs of cultivar 'Hanzhong purple'; the bulbs of cultivar 'Gailiang' had the lowest phenolic content and antioxidant capacity (FRAP, CUPRAC, MCA). The bolts of 'Hanzhong purple' also had higher phenolic content. Principal components analysis (PCA) separated the cultivars into 3 groups according to phenolic and flavonoid contents and strength of antioxidant activity. The first group had higher HRSC, FRAP, and flavonoid content; the second group had higher total phenolic content and MCA; some cultivars in the third group had higher HRSC and FRAP. All 8 test garlic bulb extracts successfully prevented Human Vascular Endothelial Cell death and significantly prevented reactive-oxygen species (ROS) formation in oxidative stress model, in which cultivar '74-x' had highest protection capability, following by cultivar 'Hanzhong purple', and the bulbs of cultivar 'No. 105 from Korea' had the lower protection capability against cell death and ROS formation. The protection capability in vivo of these garlic cultivars was consistent with their phenolic content and antioxidant capacity.
Sigma-Aldrich
Alloxazine, 96%
Pricing and availability is not currently available.