MilliporeSigma
  • Demonstration of a role for alpha-synuclein as a functional microtubule-associated protein.
Demonstration of a role for alpha-synuclein as a functional microtubule-associated protein.
Journal of Alzheimer's disease : JAD (2004-09-04)
Muhammad Abdul Alim, Qiu-Lan Ma, Kazuya Takeda, Takako Aizawa, Mamoru Matsubara, Minako Nakamura, Akiko Asada, Taro Saito, Hiroyuki Kaji, Mitsunobu Yoshii, Shinichi Hisanaga, Kenji Uéda
ABSTRACT
Alpha-synuclein is a major constituent of pathological intracellular inclusion bodies, a common feature of several neurodegenerative diseases. Two missense mutations in the alpha-synuclein gene have been identified in confirmed autosomal-dominant familial Parkinson's disease, which segregate with the illness. However, the physiological function of alpha-synuclein remains unknown. After biochemical investigations we have revealed tubulin to be an alpha-synuclein associated/binding protein. Here, we show that alpha-synuclein induces polymerization of purified tubulin into microtubules. Mutant forms of alpha-synuclein lose this potential. The binding site of alpha-synuclein to tubulin is identified, and co-localization of alpha-synuclein with microtubules is shown in cultured cells. To our knowledge, this is the first demonstration of microtubule-polymerizing activity of alpha-synuclein. Now we can see a striking resemblance between alpha-synuclein and tau: both have the same physiological function and pathological features, making abnormal structures in diseased brains known as synucleinopathies and tauopathies. The discovery of a physiological role for alpha-synuclein may provide a new dimension in researches into the mechanisms of alpha-synuclein-associated neurodegenerative diseases.
Sigma-Aldrich
α-Synuclein A53T human, recombinant, expressed in E. coli, N-terminal histidine tagged, ≥90% (SDS-PAGE), lyophilized powder
Pricing and availability is not currently available.