MilliporeSigma
  • Determination of diniconazole in agricultural samples by sol-gel immunoaffinity extraction procedure coupled with HPLC and ELISA.
Determination of diniconazole in agricultural samples by sol-gel immunoaffinity extraction procedure coupled with HPLC and ELISA.
PloS one (2012-10-12)
Zhenjiang Liu, Yahui Jin, Minghua Wang
ABSTRACT
In the European Union (EU), the use of diniconazole-M is no longer authorized. However, residues of diniconazole-M occur in various plant commodities. A selective and simple analytical method for the trace level determination of diniconazole in soil, fruit, vegetables and water samples was developed based on immunoaffinity extraction followed by Enzyme-linked immunosorbent assay (ELISA) and the high-performance liquid chromatography (HPLC) analysis. The ELISA was based on monoclonal antibodies highly specific to diniconazole and was a fast, cost-effective, and selective screening method for the detection of diniconazole. The results of the ELISA correlated well with gas chromatography (GC) results, with the correlation coefficient of 0.9879 (n = 19). A simple gel permeation chromato- graphy clean-up method was developed to purify extracts from matrices containing high amounts of fat and natural pigments, without the need for a large dilution of the sample. The immunoaffinity column (IAC) capacity was 0.180 mg g(-1). The columns could be re-used approximately 20 times with no significant alteration in capacity. The recoveries from complex samples were in the range of 89.2% to 96.1% with a relative standard deviation (RSD) of 0.770%-6.11% by ELISA. The results were in good agreement with those obtained by HPLC method. The IAC extraction procedure coupled with HPLC and ELISA analysis could be also used as alternative effective analytical methods for the determination of diniconazole concentrations in complex samples.