MilliporeSigma
NIS/TFA: a general method for hydrolyzing thioglycosides.
Carbohydrate research (2006-04-06)
Jasper Dinkelaar, Martin D Witte, Leendert J van den Bos, Herman S Overkleeft, Gijsbert A van der Marel
ABSTRACT
A variety of thioglycosides are chemoselectively hydrolyzed to the corresponding 1-hydroxy glycosides using equimolar amounts of NIS/TFA as promoter systems.
Sigma-Aldrich
N-Iodosuccinimide, 95%
Pricing and availability is not currently available.