MilliporeSigma
  • Gold(III)-catalyzed double hydroamination of o-alkynylaniline with terminal alkynes leading to N-vinylindoles.
Gold(III)-catalyzed double hydroamination of o-alkynylaniline with terminal alkynes leading to N-vinylindoles.
Organic letters (2007-02-09)
Yuhua Zhang, James P Donahue, Chao-Jun Li
ABSTRACT
A highly efficient double-hydroamination reaction of o-alkynylanilines with terminal alkynes leading to N-alkenylindoles was developed by using gold(III) as a catalyst under neat conditions. [reaction: see text].
Sigma-Aldrich
4-Ethynylanisole, 97%
Pricing and availability is not currently available.