MilliporeSigma
  • Simple palladium(II) precatalyst for Suzuki-Miyaura couplings: efficient reactions of benzylic, aryl, heteroaryl, and vinyl coupling partners.
Simple palladium(II) precatalyst for Suzuki-Miyaura couplings: efficient reactions of benzylic, aryl, heteroaryl, and vinyl coupling partners.
Organic letters (2007-12-01)
Michael J Burns, Ian J S Fairlamb, Anant R Kapdi, Petr Sehnal, Richard J K Taylor
ABSTRACT
trans-PdBr(N-Succ)(PPh3)2 (1) is a universally effective precatalyst for Suzuki-Miyaura cross-couplings of benzylic halides with aryl- or heteroarylboronic acids. Substituted aryl halides and halogenated cyclic enones can be cross-coupled with aryl- or vinylboronic acids in excellent yields. Catalyst recycling is also demonstrated.