MilliporeSigma
  • Suprofen: the pharmacology and clinical efficacy of a new non-narcotic peripheral analgesic.
Suprofen: the pharmacology and clinical efficacy of a new non-narcotic peripheral analgesic.
Clinics in rheumatic diseases (1984-08-01)
E L Tolman, M E Rosenthale, R J Capetola, J L McGuire
ABSTRACT
Suprofen is a potent, peripherally-acting, non-narcotic analgesic agent. The mechanism of action of the compound involves inhibition of prostaglandin biosynthesis and, perhaps, direct antagonism of the peripheral, pain inducing actions of prostaglandins, bradykinin and other pain mediators. Suprofen at a dose of 200 mg appears to be equal or greater in efficacy as an analgesic modality than those of ibuprofen, propoxyphene, naproxen and diflunisal or a combination of 650 mg aspirin plus 60 mg codeine. Its clinical utility has been amply demonstrated in the treatment of a number of types of pain including general and orthopedic surgery, episiotomy, post-partum pain, dysmenorrhea, dental pain and musculoskeletal disorders. Suprofen represents a new class of orally effective nonnarcotic analgesics with potential for effective clinical use in the treatment of pain.