MilliporeSigma
  • Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate: a reagent for formation of interchain carboxylic anhydrides on self-assembled monolayers.
Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate: a reagent for formation of interchain carboxylic anhydrides on self-assembled monolayers.
Langmuir : the ACS journal of surfaces and colloids (2006-07-26)
Young Shik Chi, Insung S Choi
ABSTRACT
In this paper, we report the reactivity of fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate (TFFH), a reagent for transformation of carboxylic acids into acid fluorides in solution, toward self-assembled monolayers (SAMs) of 16-mercaptohexadecanoic acid on gold. Contrary to the solution-based reactions, we found that only interchain carboxylic anhydrides (ICAs), not acid fluorides (AFs), were obtained at surfaces by the facile interchain reaction under most reaction conditions studied. AFs were found to be formed only when tetrabutylammonium fluoride, a reagent inducing fast decomposition of ICAs, was added to the reaction mixture. The reactivity of TFFH toward carboxylic acid-terminated SAMs was different from that of cyanuric fluoride, which has been reported previously (Langmuir 2005, 21, 11765-11772). This study provides more insight into the role of the proximity effect in SAM-based reactions as well as another approach to the formation of ICAs from carboxylic acid-terminated SAMs.
Sigma-Aldrich
16-Mercaptohexadecanoic acid, 98%
Pricing and availability is not currently available.