MilliporeSigma
  • Conformational variation between allelic variants of cell-surface ovine prion protein.
Conformational variation between allelic variants of cell-surface ovine prion protein.
The Biochemical journal (2004-04-09)
Alana M Thackray, Sujeong Yang, Edmond Wong, Tim J Fitzmaurice, Robert J Morgan-Warren, Raymond Bujdoso
ABSTRACT
The distribution of prion infectivity and PrPSc between peripheral lymphoid tissues suggests their possible haematogenic spread during the progression of natural scrapie in susceptible sheep. Since ovine PBMCs (peripheral blood mononuclear cells) express PrPC, they have the potential to carry or harbour disease-associated forms of PrP. To detect the possible presence of disease-associated PrP on the surface of blood cells, an understanding is required of the conformations that normal ovine cell-surface PrPC may adopt. In the present study, we have used monoclonal antibodies that recognize epitopes in either the N- or C-terminal portions of PrP to probe the conformations of PrPC on ovine PBMCs by flow cytometry. Although PBMCs from scrapie-susceptible and -resistant genotypes of sheep expressed similar levels of cell-surface PrPC, as judged by their reactivity with N-terminal-specific anti-PrP monoclonal antibodies, there was considerable genotypic heterogeneity in the region between helix-1 and residue 171. Cells from PrP-VRQ (V136R154Q171) sheep showed uniform reactivity with monoclonal antibodies that bound to epitopes around helix-1, whereas cells from PrP-ARQ (A136R154Q171) and PrP-ARR (A136R154R171) sheep showed variable binding. The region between b-strand-2 and residue 171, which includes a YYR motif, was buried or obscured in cell-surface PrPC on PBMCs from scrapie-susceptible and -resistant sheep. However, an epitope of PrPC that is influenced by residue 171 was more exposed on PBMCs from PrP-VRQ sheep than on PBMCs from the PrP-ARQ genotype. Our results highlight conformational variation between scrapie-susceptible and -resistant forms of cell-surface PrPC and also between allelic variants of susceptible genotypes.
Sigma-Aldrich
Avidin–Alkaline Phosphatase, buffered aqueous solution
Pricing and availability is not currently available.