MilliporeSigma
  • ASH2L regulates ubiquitylation signaling to MLL: trans-regulation of H3 K4 methylation in higher eukaryotes.
ASH2L regulates ubiquitylation signaling to MLL: trans-regulation of H3 K4 methylation in higher eukaryotes.
Molecular cell (2013-03-05)
Lipeng Wu, Shirley Y Lee, Bo Zhou, Uyen T T Nguyen, Tom W Muir, Song Tan, Yali Dou
ABSTRACT
Crosstalk between H2B ubiquitylation (H2Bub) and H3 K4 methylation plays important roles in coordinating functions of diverse cofactors during transcription activation. The underlying mechanism for this trans-tail signaling pathway is poorly defined in higher eukaryotes. Here, we show the following: (1) ASH2L in the MLL complex is essential for H2Bub-dependent H3 K4 methylation. Deleting or mutating K99 of the N-terminal winged helix (WH) motif in ASH2L abrogates H2Bub-dependent regulation. (2) Crosstalk can occur in trans and does not require ubiquitin to be on nucleosomes or histones to exert regulatory effects. (3) trans-regulation by ubiquitin promotes MLL activity for all three methylation states. (4) MLL3, an MLL homolog, does not respond to H2Bub, highlighting regulatory specificity for MLL family histone methyltransferases. Altogether, our results potentially expand the classic histone crosstalk to nonhistone proteins, which broadens the scope of chromatin regulation by ubiquitylation signaling.
Sigma-Aldrich
Anti-Actin antibody, Mouse monoclonal, clone AC-40, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
ChIPAb+ Histone H4 - ChIP Validated Antibody and Primer Set, rabbit monoclonal, from rabbit
Pricing and availability is not currently available.