MilliporeSigma
  • Defects in MAP1S-mediated autophagy turnover of fibronectin cause renal fibrosis.
Defects in MAP1S-mediated autophagy turnover of fibronectin cause renal fibrosis.
Aging (2016-05-30)
Guibin Xu, Fei Yue, Hai Huang, Yongzhong He, Xun Li, Haibo Zhao, Zhengming Su, Xianhan Jiang, Wenjiao Li, Jing Zou, Qi Chen, Leyuan Liu
ABSTRACT
Excessive deposition of extracellular matrix proteins in renal tissues causes renal fibrosis and renal function failure. Mammalian cells primarily use the autophagy-lysosome system to degrade misfolded/aggregated proteins and dysfunctional organelles. MAP1S is an autophagy activator and promotes the biogenesis and degradation of autophagosomes. Previously, we reported that MAP1S suppresses hepatocellular carcinogenesis in a mouse model and predicts a better prognosis in patients suffering from clear cell renal cell carcinomas. Furthermore, we have characterized that MAP1S enhances the turnover of fibronectin, and mice overexpressing LC3 but with MAP1S deleted accumulate fibronectin and develop liver fibrosis because of the synergistic impact of LC3-induced over-synthesis of fibronectin and MAP1S depletion-caused impairment of fibronectin degradation. Here we show that a suppression of MAP1S in renal cells caused an impairment of autophagy clearance of fibronectin and an activation of pyroptosis. Depletion of MAP1S in mice leads to an accumulation of fibrosis-related proteins and the development of renal fibrosis in aged mice. The levels of MAP1S were dramatically reduced and levels of fibronectin were greatly elevated in renal fibrotic tissues from patients diagnosed as renal atrophy and renal failure. Therefore, MAP1S deficiency may cause the accumulation of fibronectin and the development of renal fibrosis.
Sigma-Aldrich
Anti-Caspase-1 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.