MilliporeSigma
  • Pathological findings in a Dachshund-cross dog with neuroaxonal dystrophy.
Pathological findings in a Dachshund-cross dog with neuroaxonal dystrophy.
Acta veterinaria Scandinavica (2016-06-09)
Davide Pintus, Maria Giovanna Cancedda, Simona Macciocu, Claudia Contu, Ciriaco Ligios
ABSTRACT
Neuroaxonal dystrophy (NAD) is a neurodegenerative condition affecting humans and animals characterized by the widespread presence of swollen axons (spheroids). Herein, we report the pathological findings in a case of neuroaxonal dystrophy in a Dachshund-cross puppy, which was euthanized because of a proprioceptive positioning deficits and irreversible ataxia of the hind limbs. Histologically, there was a bilaterally symmetric neuroaxonal dystrophy with eosinophilic axonal spheroids exclusively localized at the level of the ventral posterior lateral nucleus of the thalamus, medial lemniscus, gracilis nucleus, medial cuneatus nucleus in the brain as well as the gracilis and cuneatus fasciculi throughout the spinal cord. To the authors' knowledge, this is the first report of canine neuroaxonal dystrophy with this exclusive and specific localization only in the neuronal circuit implicated in the transmission of conscious proprioceptive information.
Sigma-Aldrich
Monoclonal Anti-τ (Tau) antibody produced in mouse, clone TAU-2, ascites fluid
Pricing and availability is not currently available.