MilliporeSigma
  • Use of dynasore, the small molecule inhibitor of dynamin, in the regulation of endocytosis.
Use of dynasore, the small molecule inhibitor of dynamin, in the regulation of endocytosis.
Methods in enzymology (2008-04-17)
Tom Kirchhausen, Eric Macia, Henry E Pelish
ABSTRACT
The large GTPase dynamin is essential for clathrin-dependent coated-vesicle formation. Dynasore is a cell-permeable small molecule that inhibits the GTPase activity of dynamin1, dynamin2 and Drp1, the mitochondrial dynamin. Dynasore was discovered in a screen of approximately 16,000 compounds for inhibitors of the dynamin2 GTPase. Dynasore is a noncompetitive inhibitor of dynamin GTPase activity and blocks dynamin-dependent endocytosis in cells, including neurons. It is fast acting (seconds) and its inhibitory effect in cells can be reversed by washout. Here we present a detailed synthesis protocol for dynasore, and describe a series of experiments used to analyze the inhibitory effects of dynasore on dynamin in vitro and to study the effects of dynasore on endocytosis in cells.
Sigma-Aldrich
Dynasore hydrate
Pricing and availability is not currently available.