MilliporeSigma
  • Studies on bovine adrenal estrogen sulfotransferase. III. Facile synthesis of 3'-phospho- and 2'-phosphoadenosine 5'-phosphosulfate.
Studies on bovine adrenal estrogen sulfotransferase. III. Facile synthesis of 3'-phospho- and 2'-phosphoadenosine 5'-phosphosulfate.
Biochimica et biophysica acta (1977-02-09)
J P Horwitz, J P Neenan, R S Misra, J Rozhin, A Huo, K D Philips
PMID189816
ABSTRACT
A practical synthesis of 3'-phosphoadenosine 5'-phosphosulfate (IV) in yields of 68-72% from adenosine 2',3'-cyclic phosphate 5'-phosphate (II) is described. Reaction of II with triethylamine-N-sulfonic acid affords adenosine 2',3'-cyclic phosphate 5'-phosphosulfate (III) which, on treatment with ribonuclease-T2, provides IV. Spleen phosphodiesterase, on the other hand, converts III to 2'-phosphoadenosine 5'-phosphosulfate (V). The biological activity of IV, measured by sulfate transfer to [6,7-3H2]estrone as mediated by bovine adrenal estrone sulfotransferase (3'-phosphoadenylyl-sulfate:estrone 3-sulfotransferase, EC 2.8.2.4), is identical with that obtained with a sample of IV prepared by an established biochemical procedure. By contrast, V exhibits approximately one-third the activity of the natural isomer.
Sigma-Aldrich
Adenosine 3′-phosphate 5′-phosphosulfate lithium salt hydrate, ≥60%
Pricing and availability is not currently available.