- miR-324-3p suppresses migration and invasion by targeting WNT2B in nasopharyngeal carcinoma.
miR-324-3p suppresses migration and invasion by targeting WNT2B in nasopharyngeal carcinoma.
Cancer cell international (2017-01-06)
Chao Liu, Guo Li, Nianting Yang, Zhongwu Su, Shuiting Zhang, Tengbo Deng, Shuling Ren, Shanhong Lu, Yongquan Tian, Yong Liu, Yuanzheng Qiu
PMID28053597
ABSTRACT
Nasopharyngeal carcinoma (NPC) is a malignant epithelial carcinoma of the head and neck with strong ability of invasion and metastasis. Our previous study indicated that miR-324-3p, as a tumor-suppressive factor, could regulate radioresistance of NPC cells by targeting Quantitative real time PCR was applied to measure the expression level of miR-324-3p and Ectopic expression of miR-324-3p or silencing its target gene miR-324-3p could suppress the migration and invasion of NPC by targeting